Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Butylaniline, 97%

Click to view available options
Quantity:
25g
100g
500g
104-13-2
C10H15N
149.237
MFCD00007925
OGIQUQKNJJTLSZ-UHFFFAOYSA-N
4-n-butylaniline,benzenamine, 4-butyl,p-n-butylaniline,p-butylaniline,4-butylbenzenamine,p-aminobutylbenzene,p-butylaminobenzene,1-amino-4-butylbenzene,p-butyl aniline,4-butylphenylamine
7694
4-butylaniline
CCCCC1=CC=C(C=C1)N
104-13-2
C10H15N
149.237
MFCD00007925
OGIQUQKNJJTLSZ-UHFFFAOYSA-N
4-n-butylaniline,benzenamine, 4-butyl,p-n-butylaniline,p-butylaniline,4-butylbenzenamine,p-aminobutylbenzene,p-butylaminobenzene,1-amino-4-butylbenzene,p-butyl aniline,4-butylphenylamine
7694
4-butylaniline
CCCCC1=CC=C(C=C1)N
Flash Point 101°C (213°F)
Boiling Point 250°C
Quantity 500g
Melting Point -14°C
Beilstein 1447268
Formula Weight 149.23
Percent Purity 97%
Chemical Name or Material 4-n-Butylaniline

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : BW9470000

TSCA : TSCA