Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Butylbenzenesulfonamide, 97+%

Click to view available options
Quantity:
1g
5g
25g
1135-00-8
C10H15NO2S
213.295
MFCD00465202
IWEMSJAOIKQBBZ-UHFFFAOYSA-N
4-butylbenzene-1-sulfonamide,4-n-butylbenzenesulphonamide,4-n-butylbenzenesulfonamide,p-butylbenzenesulfonamide,4-butyl-benzenesulfonamide,maybridge1_000070,cambridge id 5152080,4-but-1-yl benzenesulphonamide,6yp
2054776
4-butylbenzenesulfonamide
CCCCC1=CC=C(C=C1)S(=O)(=O)N
1135-00-8
C10H15NO2S
213.295
MFCD00465202
IWEMSJAOIKQBBZ-UHFFFAOYSA-N
4-butylbenzene-1-sulfonamide,4-n-butylbenzenesulphonamide,4-n-butylbenzenesulfonamide,p-butylbenzenesulfonamide,4-butyl-benzenesulfonamide,maybridge1_000070,cambridge id 5152080,4-but-1-yl benzenesulphonamide,6yp
2054776
4-butylbenzenesulfonamide
CCCCC1=CC=C(C=C1)S(=O)(=O)N
Melting Point 93°C to 95°C
Quantity 5g
Formula Weight 213.29
Percent Purity ≥97%
Chemical Name or Material 4-n-Butylbenzenesulfonamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning