Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Butylbenzoic acid, 99%

Click to view available options
Quantity:
5g
25g
100g
20651-71-2
C11H14O2
178.231
MFCD00002571
JFKUBRAOUZEZSL-UHFFFAOYSA-N
4-n-butylbenzoic acid,benzoic acid, 4-butyl,4-butyl-benzoic acid,p-butylbenzoic acid,p-n-butylbenzoic acid,p-butyl benzoic acid,4-butylbenzoicacid,p-n-butyl benzoic acid,acmc-209fcp,4-n-butyl-benzoic acid
88631
4-butylbenzoic acid
CCCCC1=CC=C(C=C1)C(=O)O
20651-71-2
C11H14O2
178.231
MFCD00002571
JFKUBRAOUZEZSL-UHFFFAOYSA-N
4-n-butylbenzoic acid,benzoic acid, 4-butyl,4-butyl-benzoic acid,p-butylbenzoic acid,p-n-butylbenzoic acid,p-butyl benzoic acid,4-butylbenzoicacid,p-n-butyl benzoic acid,acmc-209fcp,4-n-butyl-benzoic acid
88631
4-butylbenzoic acid
CCCCC1=CC=C(C=C1)C(=O)O
Quantity 25g
Melting Point 100°C to 113°C
Formula Weight 178.23
Percent Purity 99%
Chemical Name or Material 4-n-Butylbenzoic acid

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 243-940-9

TSCA : Yes

Recommended Storage : Ambient temperatures