Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Butylbenzonitrile, 98%

Click to view available options
Quantity:
5g
25g
100g
20651-73-4
C11H13N
159.232
MFCD00052345
KGZNJCXNPLUEQS-UHFFFAOYSA-N
4-n-butylbenzonitrile,benzonitrile,4-butyl,benzonitrile, 4-butyl,4-butylbenzenecarbonitrile,acmc-20ammb,#,1-butyl-4-cyanobenzene,4-n-butyl benzonitrile,4-butylbenzonitrile
140730
4-butylbenzonitrile
CCCCC1=CC=C(C=C1)C#N
20651-73-4
C11H13N
159.232
MFCD00052345
KGZNJCXNPLUEQS-UHFFFAOYSA-N
4-n-butylbenzonitrile,benzonitrile,4-butyl,benzonitrile, 4-butyl,4-butylbenzenecarbonitrile,acmc-20ammb,#,1-butyl-4-cyanobenzene,4-n-butyl benzonitrile,4-butylbenzonitrile
140730
4-butylbenzonitrile
CCCCC1=CC=C(C=C1)C#N
Density 0.924
Quantity 100g
Odor Almond-like
Boiling Point 130°C to 132°C (15mmHg)
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzonitrile

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures