Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Butylbenzylamine, 98%

Click to view available options
Quantity:
5g
25g
100g
57802-79-6
C11H17N
163.264
MFCD02258855
IBVGSPOHLFKLHM-UHFFFAOYSA-N
4-butylbenzylamine,4-n-butylbenzylamine,4-butylphenyl methanamine,4-butylphenyl methylamine,1-4-butylphenyl methanamine,p-n-butylbenzylamin,4-butyl-benzylamine,4-butylbenzenemethanamine,benzenemethanamine, 4-butyl
4029871
(4-butylphenyl)methanamine
CCCCC1=CC=C(C=C1)CN
57802-79-6
C11H17N
163.264
MFCD02258855
IBVGSPOHLFKLHM-UHFFFAOYSA-N
4-butylbenzylamine,4-n-butylbenzylamine,4-butylphenyl methanamine,4-butylphenyl methylamine,1-4-butylphenyl methanamine,p-n-butylbenzylamin,4-butyl-benzylamine,4-butylbenzenemethanamine,benzenemethanamine, 4-butyl
4029871
(4-butylphenyl)methanamine
CCCCC1=CC=C(C=C1)CN
Sensitivity Air Sensitive
Quantity 100g
Boiling Point 89°C to 91°C (1mmHg)
UN Number UN2735
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Butylbenzylamine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon