Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(N-Cyclopentylcarbamoyl)benzeneboronic acid pinacol ester, 95%

Manufacturer:  Alfa Aesar™ H5226906

Quantity 5g

 View more versions of this product

Catalog No. AAH5226906


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

933987-10-1
315.22
N-Cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
C18H26BNO3
MFCD16660857
4-(N-Cyclopentylcarbamoyl)benzeneboronic acid pinacol ester

Specifications

933987-10-1
315.22
5g
4-(N-Cyclopentylcarbamoyl)benzeneboronic acid pinacol ester
C18H26BNO3
MFCD16660857
N-Cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
95%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit