Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Decylbiphenyl, 98%

Click to view available options
Quantity:
1g
5g
93972-02-2
C22H30
294.482
MFCD00102117
YYBDFGOLQQVHDV-UHFFFAOYSA-N
4-n-decylbiphenyl,4-decyl-biphenyl,4-decyl-1,1'-biphenyl,4-decylbiphenyl,4-decyl-1-phenylbenzene,maybridge1_002181,1,1'-biphenyl, 4-decyl
2801612
1-decyl-4-phenylbenzene
CCCCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2
93972-02-2
C22H30
294.482
MFCD00102117
YYBDFGOLQQVHDV-UHFFFAOYSA-N
4-n-decylbiphenyl,4-decyl-biphenyl,4-decyl-1,1'-biphenyl,4-decylbiphenyl,4-decyl-1-phenylbenzene,maybridge1_002181,1,1'-biphenyl, 4-decyl
2801612
1-decyl-4-phenylbenzene
CCCCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2
Applications
PDLC monomer. As chemical reagent.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Boiling Point 204°C to 210°C (1.5mmHg)
Melting Point 52°C to 54°C
Quantity 1g
Solubility Information Insoluble in water.
Formula Weight 294.48
Percent Purity 98%
Chemical Name or Material 4-n-Decylbiphenyl

TSCA : No

Recommended Storage : Ambient temperatures