Alfa Aesar™ 4-n-Heptylaniline, 98%

Manufacturer: Alfa Aesar™ H2679303

Quantity 1g

 View more versions of this product

Catalog No. AAH2679303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37529-27-4
    Molecular Formula C13H21N
    Formula Weight 191.31
    MDL Number MFCD00007928
    Chemical Name or Material 4-n-Heptylaniline

    Specifications

    Formula Weight 191.31
    CAS 37529-27-4
    Health Hazard 2 GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    Flash Point >110°C (230°F)
    Quantity 1g
    Molecular Formula C13H21N
    MDL Number MFCD00007928
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 159°C (18mmHg)
    Chemical Name or Material 4-n-Heptylaniline
    Assay 98%

    GHS H Statement
    H301-H315-H319
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280-P301+P310-P305+P351+P338-P362-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Take off contaminated clothing and wash before reuse.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit