Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Heptylbenzoic acid, 99+%

Click to view available options
Quantity:
5g
25g
38350-87-7
C14H20O2
220.312
MFCD00009722
VSUKEWPHURLYTK-UHFFFAOYSA-N
benzoic acid, 4-heptyl,4-n-heptylbenzoic acid,4-heptylbenzoicacid,4-heptylbenzoate,p-heptylbenzoic acid,acmc-209izm,4-n-heptyl-benzoic acid,4-heptylbenzoic acid
170036
4-heptylbenzoic acid
CCCCCCCC1=CC=C(C=C1)C(=O)O
38350-87-7
C14H20O2
220.312
MFCD00009722
VSUKEWPHURLYTK-UHFFFAOYSA-N
benzoic acid, 4-heptyl,4-n-heptylbenzoic acid,4-heptylbenzoicacid,4-heptylbenzoate,p-heptylbenzoic acid,acmc-209izm,4-n-heptyl-benzoic acid,4-heptylbenzoic acid
170036
4-heptylbenzoic acid
CCCCCCCC1=CC=C(C=C1)C(=O)O
Quantity 5g
Melting Point 101°C to 102°C
Beilstein 2047135
Formula Weight 220.31
Percent Purity ≥99%
Chemical Name or Material 4-n-Heptylbenzoic acid

P261-P264b-P270-P271-P280i-P301+P312-P304+P340-P305+P351+P338-P312-P330-P501c

H302-H319-H335

EINECSNumber : 253-894-1

TSCA : Yes

Recommended Storage : Ambient temperatures