Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Hexylbenzeneboronic acid, 97%

Click to view available options
Quantity:
250mg
1g
105365-50-2
C12H19BO2
206.092
MFCD04039029
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
4-hexylphenyl boronic acid,4-n-hexylphenylboronic acid,4-hex-1-yl benzeneboronic acid,1-borono-4-hex-1-yl benzene,4-n-hexylbenzeneboronic acid,boronic acid, 4-hexylphenyl,4-hexylphenylboronicacid,p-hexylphenylboronic acid,4-hexylbenzeneboronic acid,4-hexylphenyl-boronic acid
4197864
(4-hexylphenyl)boronic acid
B(C1=CC=C(C=C1)CCCCCC)(O)O
105365-50-2
C12H19BO2
206.092
MFCD04039029
CFYBKGAFWFRFRJ-UHFFFAOYSA-N
4-hexylphenyl boronic acid,4-n-hexylphenylboronic acid,4-hex-1-yl benzeneboronic acid,1-borono-4-hex-1-yl benzene,4-n-hexylbenzeneboronic acid,boronic acid, 4-hexylphenyl,4-hexylphenylboronicacid,p-hexylphenylboronic acid,4-hexylbenzeneboronic acid,4-hexylphenyl-boronic acid
4197864
(4-hexylphenyl)boronic acid
B(C1=CC=C(C=C1)CCCCCC)(O)O
Quantity 250mg
Formula Weight 206.09
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 4-n-Hexylbenzeneboronic acid