Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Hexylbenzoyl chloride, 98%

Click to view available options
Quantity:
5g
25g
100g
50606-95-6
C13H17ClO
224.728
MFCD00000701
XRAHLPNMIIAEPP-UHFFFAOYSA-N
4-n-hexylbenzoyl chloride,benzoyl chloride, 4-hexyl,p-hexylbenzoyl chloride,4-n-hexylbenzoylchloride,4-n-hexyl-benzoyl chloride,4-hexylbenzoyl chloride,para-n-hexyl benzoic acid chloride,4-hexylbenzene-1-carbonyl chloride
170863
4-hexylbenzoyl chloride
CCCCCCC1=CC=C(C=C1)C(=O)Cl
50606-95-6
C13H17ClO
224.728
MFCD00000701
XRAHLPNMIIAEPP-UHFFFAOYSA-N
4-n-hexylbenzoyl chloride,benzoyl chloride, 4-hexyl,p-hexylbenzoyl chloride,4-n-hexylbenzoylchloride,4-n-hexyl-benzoyl chloride,4-hexylbenzoyl chloride,para-n-hexyl benzoic acid chloride,4-hexylbenzene-1-carbonyl chloride
170863
4-hexylbenzoyl chloride
CCCCCCC1=CC=C(C=C1)C(=O)Cl
Refractive Index 1.5256
Sensitivity Moisture sensitive
Density 1.029
Flash Point >110°C (230°F)
Boiling Point 176°C to 177°C (22mmHg)
Quantity 100g
UN Number UN3265
Beilstein 2208938
Formula Weight 224.73
Percent Purity 98%
Chemical Name or Material 4-n-Hexylbenzoyl chloride
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 256-647-6

TSCA : Yes

Recommended Storage : Ambient temperatures