Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(N,N-Dimethylamino)phenylboronic acid, 96%, Acros Organics

Click to view available options
Quantity:
500mg
2.5g
Packaging:
Glass bottle
28611-39-4
C8H12BNO2
165
MFCD01074642
RIIPFHVHLXPMHQ-UHFFFAOYSA-N
4-dimethylamino phenylboronic acid,4-dimethylamino phenyl boronic acid,4-n,n-dimethylamino phenylboronic acid,4-dimethylamino benzeneboronic acid,4-dimethylaminophenylboronic acid,4-dimethylaminophenyl boronic acid,4-dimethylamino phenyl boranediol,4-n,n-dimethylphenylboronic acid
2734344
[4-(dimethylamino)phenyl]boronic acid
B(C1=CC=C(C=C1)N(C)C)(O)O
28611-39-4
C8H12BNO2
165
MFCD01074642
RIIPFHVHLXPMHQ-UHFFFAOYSA-N
4-dimethylamino phenylboronic acid,4-dimethylamino phenyl boronic acid,4-n,n-dimethylamino phenylboronic acid,4-dimethylamino benzeneboronic acid,4-dimethylaminophenylboronic acid,4-dimethylaminophenyl boronic acid,4-dimethylamino phenyl boranediol,4-n,n-dimethylphenylboronic acid
2734344
[4-(dimethylamino)phenyl]boronic acid
B(C1=CC=C(C=C1)N(C)C)(O)O
Melting Point 227°C
Color Blue-Gray or Gray-Green to White
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 2.5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 96%
Formula Weight 165
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 4-(N, N-Dimethylamino)phenylboronic acid
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

GHS Signal Word: Warning