Alfa Aesar™ 4-n-Octylaniline, 99%

Manufacturer: Alfa Aesar™ H2671303

Quantity 1g

 View more versions of this product

Catalog No. AAH2671303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16245-79-7
    Molecular Formula C14H23N
    Formula Weight 205.34
    MDL Number MFCD00007929
    Chemical Name or Material 4-n-Octylaniline

    Specifications

    Formula Weight 205.34
    Boiling Point 175°C (13mmHg)
    Quantity 1g
    Flash Point 110°C (230°F)
    Molecular Formula C14H23N
    MDL Number MFCD00007929
    CAS 16245-79-7
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.
    Health Hazard 3 GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 4-n-Octylaniline
    Assay 99%

    Description

    Applications
    4-Octylaniline can be been used as an extractant during the extraction of microgram level concentrations of ruthenium(IV) from halide medium. It can also been used in the synthesis of selective benzimidazole based analogs of sphingosine-1-phosphate.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H301-H311
    Toxic if swallowed.
    Toxic in contact with skin.

    GHS P Statement
    P280-P301+P310-P361-P312-P405-P501a
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    Remove/Take off immediately all contaminated clothing.
    Call a POISON CENTER or doctor/physician if you feel unwell.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit