Alfa Aesar™ 4-n-Octylbiphenyl, 97%

Manufacturer: Alfa Aesar™ B2501006

Quantity 5g

 View more versions of this product

Catalog No. AAB2501006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7116-97-4
    Molecular Formula C20H26
    Formula Weight 266.43
    MDL Number MFCD00102113
    Chemical Name or Material 4-n-Octylbiphenyl

    Specifications

    Formula Weight 266.43
    Quantity 5g
    Molecular Formula C20H26
    MDL Number MFCD00102113
    CAS 7116-97-4
    Chemical Name or Material 4-n-Octylbiphenyl
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit