Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Octylbiphenyl, 97%

Click to view available options
Quantity:
1g
5g
25g
7116-97-4
C20H26
266.428
MFCD00102113
KCLBLDDJJJNGBC-UHFFFAOYSA-N
4-octylbiphenyl,4-octyl-1,1'-biphenyl,4-n-octylbiphenyl,4-octyldiphenyl,1,1'-biphenyl, 4-octyl,biphenyl, 4-octyl-6ci,7ci,8ci,4-octyl-1-phenylbenzene,4-05-00-02003 beilstein handbook reference
201918
1-octyl-4-phenylbenzene
CCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2
7116-97-4
C20H26
266.428
MFCD00102113
KCLBLDDJJJNGBC-UHFFFAOYSA-N
4-octylbiphenyl,4-octyl-1,1'-biphenyl,4-n-octylbiphenyl,4-octyldiphenyl,1,1'-biphenyl, 4-octyl,biphenyl, 4-octyl-6ci,7ci,8ci,4-octyl-1-phenylbenzene,4-05-00-02003 beilstein handbook reference
201918
1-octyl-4-phenylbenzene
CCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2
Quantity 25g
Formula Weight 266.43
Percent Purity 97%
Chemical Name or Material 4-n-Octylbiphenyl