Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(n-Octyloxy)phenol, 98%

Click to view available options
Quantity:
1g
5g
3780-50-5
C14H22O2
222.328
MFCD00045779
HFRUPPHPJRZOCM-UHFFFAOYSA-N
4-octyloxy phenol,4-n-octyloxy phenol,4-n-octyloxyphenol,phenol, 4-octyloxy,4-octyloxyphenol,p-octyloxyphenol,p-octyloxy phenol,4-n-octoxyphenol,pubchem13211,acmc-209ivj
77412
4-octoxyphenol
CCCCCCCCOC1=CC=C(C=C1)O
3780-50-5
C14H22O2
222.328
MFCD00045779
HFRUPPHPJRZOCM-UHFFFAOYSA-N
4-octyloxy phenol,4-n-octyloxy phenol,4-n-octyloxyphenol,phenol, 4-octyloxy,4-octyloxyphenol,p-octyloxyphenol,p-octyloxy phenol,4-n-octoxyphenol,pubchem13211,acmc-209ivj
77412
4-octoxyphenol
CCCCCCCCOC1=CC=C(C=C1)O
Quantity 5g
Melting Point 80°C
Formula Weight 222.33
Percent Purity 98%
Chemical Name or Material 4-(n-Octyloxy)phenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 223-243-6

TSCA : Yes

Recommended Storage : Ambient temperatures