Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(n-Octyloxy)phenol, 98%, Acros Organics

Click to view available options
Quantity:
5g
3780-50-5
C14H22O2
222.328
HFRUPPHPJRZOCM-UHFFFAOYSA-N
4-octyloxy phenol,4-n-octyloxy phenol,4-n-octyloxyphenol,phenol, 4-octyloxy,4-octyloxyphenol,p-octyloxyphenol,p-octyloxy phenol,4-n-octoxyphenol,pubchem13211,acmc-209ivj
77412
4-octoxyphenol
CCCCCCCCOC1=CC=C(C=C1)O
3780-50-5
C14H22O2
222.328
HFRUPPHPJRZOCM-UHFFFAOYSA-N
4-octyloxy phenol,4-n-octyloxy phenol,4-n-octyloxyphenol,phenol, 4-octyloxy,4-octyloxyphenol,p-octyloxyphenol,p-octyloxy phenol,4-n-octoxyphenol,pubchem13211,acmc-209ivj
77412
4-octoxyphenol
CCCCCCCCOC1=CC=C(C=C1)O
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 222.33
Chemical Name or Material 4-(n-Octyloxy)phenol, 98%

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-243-6

TSCA : TSCA