Alfa Aesar™ 4'-n-Octyloxyacetophenone

Manufacturer: Alfa Aesar™ H5068406

Quantity 5g

 View more versions of this product

Catalog No. AAH5068406

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 37062-63-8
    Molecular Formula C16H24O2
    Formula Weight 248.37
    MDL Number MFCD00143208
    Chemical Name or Material 4'-n-Octyloxyacetophenone

    Specifications

    Formula Weight 248.37
    Quantity 5g
    Molecular Formula C16H24O2
    MDL Number MFCD00143208
    CAS 37062-63-8
    Chemical Name or Material 4'-n-Octyloxyacetophenone

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit