Alfa Aesar™ 4-n-Octylphenol, 99%

Manufacturer: Alfa Aesar™ H2639603

Quantity 1g

 View more versions of this product

Catalog No. AAH2639603

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1806-26-4
    Molecular Formula C14H22O
    Formula Weight 206.32
    MDL Number MFCD00036134
    UN Number UN2430
    Chemical Name or Material 4-n-Octylphenol

    Specifications

    Formula Weight 206.32
    Molecular Formula C14H22O
    MDL Number MFCD00036134
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318-H361
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of damaging fertility or the unborn child.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 150°C (4mmHg)
    Flash Point >110°C (230°F)
    Quantity 1g
    RTECS Number SM5787000
    TSCA TSCA
    CAS 1806-26-4
    UN Number UN2430
    Melting Point 43° to 45°C
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
    Chemical Name or Material 4-n-Octylphenol
    Assay 99%

    Description

    Applications
    4-n-Octylphenol is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

    Solubility
    Insoluble in water.

    Notes
    Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

    GHS H Statement
    H301-H314-H318-H361
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of damaging fertility or the unborn child.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit