Alfa Aesar™ 4'-n-Pentylacetophenone, 98%

Manufacturer: Alfa Aesar™ A1072506

Quantity 5g

 View more versions of this product

Catalog No. AAA1072506

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 37593-02-5
    Molecular Formula C13H18O
    Formula Weight 190.29
    MDL Number MFCD00043687
    Synonym 4'-n-Amylacetophenone
    Chemical Name or Material 4'-n-Pentylacetophenone

    Specifications

    Formula Weight 190.29
    TSCA TSCA
    Molecular Formula C13H18O
    CAS 37593-02-5
    Synonym 4'-n-Amylacetophenone
    Quantity 5g
    MDL Number MFCD00043687
    Boiling Point 132° to 133°C (4mmHg)
    Chemical Name or Material 4'-n-Pentylacetophenone
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit