Alfa Aesar™ 4-n-Pentylbiphenyl, 99%

Manufacturer: Alfa Aesar™ A1259014

Quantity 25g

 View more versions of this product

Catalog No. AAA1259014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7116-96-3
    Molecular Formula C17H20
    Formula Weight 224.35
    MDL Number MFCD00009503
    Beilstein 2082056
    Synonym 4-n-Amylbiphenyl
    Chemical Name or Material 4-n-Pentylbiphenyl

    Specifications

    Formula Weight 224.35
    TSCA TSCA
    Molecular Formula C17H20
    MDL Number MFCD00009503
    Beilstein 2082056
    Flash Point >110°C (230°F)
    Quantity 25g
    CAS 7116-96-3
    Synonym 4-n-Amylbiphenyl
    Boiling Point 166° to 167°C (6mmHg)
    Melting Point 9° to 11°C
    Chemical Name or Material 4-n-Pentylbiphenyl
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit