Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Pentylcyclohexanone, 98+%

Click to view available options
Quantity:
10g
50g
61203-83-6
C11H20O
168.28
MFCD00059581
UKLNPJDLSPMJMQ-UHFFFAOYSA-N
4-pentylcyclohexanone,4-n-pentylcyclohexanone,4-amylcyclohexanone,4-n-amylcyclohexanone,4-pentyl-1-cyclohexanone,4-pentylcyclo hexanone,pubchem20247,acmc-209mq3,uklnpjdlspmjmq-uhfffaoysa
566097
4-pentylcyclohexan-1-one
CCCCCC1CCC(=O)CC1
61203-83-6
C11H20O
168.28
MFCD00059581
UKLNPJDLSPMJMQ-UHFFFAOYSA-N
4-pentylcyclohexanone,4-n-pentylcyclohexanone,4-amylcyclohexanone,4-n-amylcyclohexanone,4-pentyl-1-cyclohexanone,4-pentylcyclo hexanone,pubchem20247,acmc-209mq3,uklnpjdlspmjmq-uhfffaoysa
566097
4-pentylcyclohexan-1-one
CCCCCC1CCC(=O)CC1
Quantity 10g
UN Number UN3082
Formula Weight 168.28
Percent Purity ≥98%
Chemical Name or Material 4-n-Pentylcyclohexanone

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.