Alfa Aesar™ 4-(N-Phthalimidomethyl)benzeneboronic acid pinacol ester, 95%

Manufacturer: Alfa Aesar™ H5226706

Quantity 5g

 View more versions of this product

Catalog No. AAH5226706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 138500-87-5
    Molecular Formula C21H22BNO4
    Formula Weight 363.22
    MDL Number MFCD02179489
    Synonym N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]phthalimide
    Chemical Name or Material 4-(N-Phthalimidomethyl)benzeneboronic acid pinacol ester

    Specifications

    Formula Weight 363.22
    Molecular Formula C21H22BNO4
    MDL Number MFCD02179489
    CAS 138500-87-5
    Synonym N-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]phthalimide
    Quantity 5g
    Chemical Name or Material 4-(N-Phthalimidomethyl)benzeneboronic acid pinacol ester
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit