Alfa Aesar™ 4'-n-Propoxybiphenyl-4-boronic acid, 97%

Manufacturer: Alfa Aesar™ H55999MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH55999MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 849062-20-0
    Formula Weight 256.11
    MDL Number MFCD06798061
    Synonym 4-(4'-Propoxyphenyl)phenylboronic acid
    Chemical Name or Material 4'-n-Propoxybiphenyl-4-boronic acid

    Specifications

    Formula Weight 256.11
    Physical Form Solid
    MDL Number MFCD06798061
    CAS 849062-20-0
    Synonym 4-(4'-Propoxyphenyl)phenylboronic acid
    Quantity 250mg
    Chemical Name or Material 4'-n-Propoxybiphenyl-4-boronic acid
    Assay 97%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit