Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Propoxyphenol, 98%

Click to view available options
Quantity:
5g
25g
100g
18979-50-5
C9H12O2
152.193
MFCD00002335
KIIIPQXXLVCCQP-UHFFFAOYSA-N
phenol, 4-propoxy,4-n-propoxyphenol,p-propoxyphenol,phenol, p-propoxy,hydroquinone monopropyl ether,p-propoxy phenol,4-propyloxyphenol,4-propoxy-phenol,pubchem13204,para-propoxyphenol
29352
4-propoxyphenol
CCCOC1=CC=C(C=C1)O
18979-50-5
C9H12O2
152.193
MFCD00002335
KIIIPQXXLVCCQP-UHFFFAOYSA-N
phenol, 4-propoxy,4-n-propoxyphenol,p-propoxyphenol,phenol, p-propoxy,hydroquinone monopropyl ether,p-propoxy phenol,4-propyloxyphenol,4-propoxy-phenol,pubchem13204,para-propoxyphenol
29352
4-propoxyphenol
CCCOC1=CC=C(C=C1)O
Quantity 100g
Flash Point >110°C (230°F)
Melting Point 55°C to 58°C
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 4-n-Propoxyphenol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA