Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Propylphenol, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass Bottle
645-56-7
C9H12O
136.194
MFCD00002395
KLSLBUSXWBJMEC-UHFFFAOYSA-N
4-n-propylphenol,p-propylphenol,phenol, 4-propyl,dihydrochavicol,phenol, p-propyl,1-hydroxy-4-n-propylbenzene,p-n-propylphenol,4-propyl-phenol,p-hydroxypropylbenzene,hydroxypropylbenzene, p
12580
CHEBI:34434
4-propylphenol
CCCC1=CC=C(C=C1)O
645-56-7
C9H12O
136.194
MFCD00002395
KLSLBUSXWBJMEC-UHFFFAOYSA-N
4-n-propylphenol,p-propylphenol,phenol, 4-propyl,dihydrochavicol,phenol, p-propyl,1-hydroxy-4-n-propylbenzene,p-n-propylphenol,4-propyl-phenol,p-hydroxypropylbenzene,hydroxypropylbenzene, p
12580
CHEBI:34434
4-propylphenol
CCCC1=CC=C(C=C1)O
Refractive Index 1.5220 to 1.5240
Packaging Glass Bottle
Color Yellow
Flash Point 106°C
Specific Gravity 0.983
Infrared Spectrum Authentic
Quantity 1g
Density 0.9830g/mL
Melting Point 22.0°C
Boiling Point 232.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula CH3CH2CH2C6H4OH
Formula Weight 136.19
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 4-n-Propylphenol
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Do not eat, drink or smoke when using this product.
Wash face, hands and any exposed skin thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger