Alfa Aesar™ 4-n-Propylphenylacetylene, 97%

Manufacturer: Alfa Aesar™ B2506522

Quantity 100g

 View more versions of this product

Catalog No. AAB2506522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62452-73-7
    Molecular Formula C11H12
    Formula Weight 144.22
    MDL Number MFCD00173886
    Synonym 1-Ethynyl-4-n-propylbenzene
    Chemical Name or Material 4-n-Propylphenylacetylene

    Specifications

    Formula Weight 144.22
    Molecular Formula C11H12
    MDL Number MFCD00173886
    CAS 62452-73-7
    Synonym 1-Ethynyl-4-n-propylbenzene
    Health Hazard 1 Warning
    Flash Point 67°C (153°F)
    Boiling Point 205°C
    Quantity 100g
    Chemical Name or Material 4-n-Propylphenylacetylene
    Assay 97%

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit