Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-(n-Undecyloxy)benzoic acid, 98%

Manufacturer:  Alfa Aesar™ B2472703

Quantity 1g

 View more versions of this product

Catalog No. AAB2472703


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

15872-44-3
292.42
4-(n-Undecyloxy)benzoic acid
C18H28O3
MFCD00043772

Specifications

96° to 137°C
15872-44-3
292.42
4-(n-Undecyloxy)benzoic acid
1g
C18H28O3
MFCD00043772
98%
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit