Alfa Aesar™ 4-(n-Undecyloxy)benzoic acid, 98%

Manufacturer: Alfa Aesar™ B2472714

Quantity 25g

 View more versions of this product

Catalog No. AAB2472714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15872-44-3
    Molecular Formula C18H28O3
    Formula Weight 292.42
    MDL Number MFCD00043772
    Chemical Name or Material 4-(n-Undecyloxy)benzoic acid

    Specifications

    Formula Weight 292.42
    Molecular Formula C18H28O3
    MDL Number MFCD00043772
    CAS 15872-44-3
    Boiling Point 96° to 137°C
    Quantity 25g
    Chemical Name or Material 4-(n-Undecyloxy)benzoic acid
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit