Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Nitro-2-(trifluoromethyl)anisole, 98+%

Click to view available options
Quantity:
5g
25g
100g
654-76-2
C8H6F3NO3
221.135
MFCD00007328
KGFADEJSZXEVMC-UHFFFAOYSA-N
2-methoxy-5-nitrobenzotrifluoride,1-methoxy-4-nitro-2-trifluoromethyl benzene,4-nitro-2-trifluoromethyl anisole,5-nitro-2-methoxybenzotrifluoride,benzene, 1-methoxy-4-nitro-2-trifluoromethyl,1-methoxy-4-nitro-2-trifluoromethyl-benzene,2-methoxy-5-nitro trifluoromethyl benzene,pubchem4875,acmc-209nr0,2-methoxy-5-nitro-benzotrifluoride
69562
1-methoxy-4-nitro-2-(trifluoromethyl)benzene
COC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F
654-76-2
C8H6F3NO3
221.135
MFCD00007328
KGFADEJSZXEVMC-UHFFFAOYSA-N
2-methoxy-5-nitrobenzotrifluoride,1-methoxy-4-nitro-2-trifluoromethyl benzene,4-nitro-2-trifluoromethyl anisole,5-nitro-2-methoxybenzotrifluoride,benzene, 1-methoxy-4-nitro-2-trifluoromethyl,1-methoxy-4-nitro-2-trifluoromethyl-benzene,2-methoxy-5-nitro trifluoromethyl benzene,pubchem4875,acmc-209nr0,2-methoxy-5-nitro-benzotrifluoride
69562
1-methoxy-4-nitro-2-(trifluoromethyl)benzene
COC1=C(C=C(C=C1)[N+](=O)[O-])C(F)(F)F
Quantity 25g
Melting Point 79°C to 81°C
UN Number UN1759
Beilstein 2129987
Formula Weight 221.14
Chemical Name or Material 4-Nitro-2-(trifluoromethyl)anisole

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.