Alfa Aesar™ 4-Nitro-D-phenylalanine, 98%

Manufacturer: Alfa Aesar™ H26959MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH26959MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 56613-61-7
    Molecular Formula C9H10N2O4
    Formula Weight 210.19
    MDL Number MFCD00066016
    Synonym H-D-Phe(4-NO2)-OH
    Chemical Name or Material 4-Nitro-D-phenylalanine

    Specifications

    Formula Weight 210.19
    Health Hazard 1 Warning
    Quantity 250mg
    Health Hazard 3 GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C9H10N2O4
    MDL Number MFCD00066016
    CAS 56613-61-7
    Synonym H-D-Phe(4-NO2)-OH
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material 4-Nitro-D-phenylalanine
    Assay 98%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P264-P270-P301+P312-P330-P501a
    Wash thoroughly after handling.
    Do not eat, drink or smoke when using this product.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit