Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-m-xylene, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
Packaging:
Glass bottle
89-87-2
C8H9NO2
151.165
MFCD00007169
BBUPBICWUURTNP-UHFFFAOYSA-N
4-nitro-m-xylene, 1,3-dimethyl-4-nitrobenzene, 4-nitro-1,3-dimethylbenzene, 2,4-dimethylnitrobenzene, benzene, 2,4-dimethyl-1-nitro, m-xylene, 4-nitro, 4-nitro-1,3-xylene, 1-nitro-2,4-dimethylbenzene, unii-7em84eue05, ccris 3120
6991
2,4-dimethyl-1-nitrobenzene
CC1=CC(=C(C=C1)[N+](=O)[O-])C
89-87-2
C8H9NO2
151.165
MFCD00007169
BBUPBICWUURTNP-UHFFFAOYSA-N
4-nitro-m-xylene, 1,3-dimethyl-4-nitrobenzene, 4-nitro-1,3-dimethylbenzene, 2,4-dimethylnitrobenzene, benzene, 2,4-dimethyl-1-nitro, m-xylene, 4-nitro, 4-nitro-1,3-xylene, 1-nitro-2,4-dimethylbenzene, unii-7em84eue05, ccris 3120
6991
2,4-dimethyl-1-nitrobenzene
CC1=CC(=C(C=C1)[N+](=O)[O-])C
Quantity 100g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
Refractive Index 1.5485 to 1.5505 (20°C, 589nm)
Density 1.11g/mL
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)2C6H3NO2
Beilstein 05, 378
Merck Index 10, 6503
Formula Weight 151.16
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material 4-Nitro-m-xylene, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning