Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroanisole 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
250g
Packaging:
Glass bottle
100-17-4
C7H7NO3
153.14
MFCD00007327
BNUHAJGCKIQFGE-UHFFFAOYSA-N
4-nitroanisole,p-nitroanisole,4-methoxynitrobenzene,p-methoxynitrobenzene,benzene, 1-methoxy-4-nitro,nitroanisole,p-nitroanisol,anisole, p-nitro,4-nitrophenyl methyl ether,unii-g989z7wolh
7485
CHEBI:1911
1-methoxy-4-nitrobenzene
COC1=CC=C(C=C1)[N+](=O)[O-]
100-17-4
C7H7NO3
153.14
MFCD00007327
BNUHAJGCKIQFGE-UHFFFAOYSA-N
4-nitroanisole,p-nitroanisole,4-methoxynitrobenzene,p-methoxynitrobenzene,benzene, 1-methoxy-4-nitro,nitroanisole,p-nitroanisol,anisole, p-nitro,4-nitrophenyl methyl ether,unii-g989z7wolh
7485
CHEBI:1911
1-methoxy-4-nitrobenzene
COC1=CC=C(C=C1)[N+](=O)[O-]
Melting Point 51.0°C to 53.0°C
Packaging Glass bottle
Specific Gravity 1.23
Boiling Point 260.0°C
Infrared Spectrum Authentic
Quantity 5g
Flash Point 130°C
Density 1.2300g/mL
Color Brown to Green
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula O2NC6H4OCH3
Beilstein 06, 230
Merck Index 15, 6671
Solubility Information Solubility in water: 0.468g/L (20°C). Other solubilities: soluble in alcohol and ether
Formula Weight 153.14
Physical Form Crystals
Percent Purity ≥99%
Chemical Name or Material 4-Nitroanisole, 99+%
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-825-3

RTECSNumber : BZ8800000

TSCA : TSCA