Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzaldoxime, 98%, Thermo Scientific Chemicals

Catalog No. AAA1552514
Click to view available options
Quantity:
25 g

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1129-37-9
Molecular Formula C7H6N2O3
Molecular Weight (g/mol) 166.14
MDL Number MFCD00007377 MFCD00007377 MFCD00151329 MFCD00151329 MFCD00165078
InChI Key WTLPAVBACRIHHC-VMPITWQZSA-N
Synonym syn-4-nitrobenzaldoxime, 4-nitrobenzaldehyde oxime, p-nitrobenzaldoxime, anti-p-nitrobenzaldoxime, p-nitro-anti-benzaldoxime, wln: wnr d1unq, e-4-nitrobenzaldehyde oxime, 4-nitrobenzaldehyde oxime, cis, 1z-4-nitrobenzaldehyde oxime, benzaldehyde, p-nitro-, oxime, z
PubChem CID 5374047
SMILES O\N=C\C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 126°C to 130°C
Quantity 25 g
UN Number UN2811
Beilstein 973581
IUPAC Name (NZ)-N-[(4-nitrophenyl)methylidene]hydroxylamine
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzaldoxime
Hazard Category H301-H315-H319-H332-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 214-445-5
RTECSNumber CU7450000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.