Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Nitrobenzaldoxime, 98%

Click to view available options
Quantity:
5g
25g
1129-37-9
C7H6N2O3
166.136
MFCD00007377
WTLPAVBACRIHHC-YVMONPNESA-N
syn-4-nitrobenzaldoxime,4-nitrobenzaldehyde oxime,p-nitrobenzaldoxime,anti-p-nitrobenzaldoxime,p-nitro-anti-benzaldoxime,wln: wnr d1unq,e-4-nitrobenzaldehyde oxime,4-nitrobenzaldehyde oxime, cis,1z-4-nitrobenzaldehyde oxime,benzaldehyde, p-nitro-, oxime, z
5374047
(NZ)-N-[(4-nitrophenyl)methylidene]hydroxylamine
C1=CC(=CC=C1C=NO)[N+](=O)[O-]
1129-37-9
C7H6N2O3
166.136
MFCD00007377
WTLPAVBACRIHHC-YVMONPNESA-N
syn-4-nitrobenzaldoxime,4-nitrobenzaldehyde oxime,p-nitrobenzaldoxime,anti-p-nitrobenzaldoxime,p-nitro-anti-benzaldoxime,wln: wnr d1unq,e-4-nitrobenzaldehyde oxime,4-nitrobenzaldehyde oxime, cis,1z-4-nitrobenzaldehyde oxime,benzaldehyde, p-nitro-, oxime, z
5374047
(NZ)-N-[(4-nitrophenyl)methylidene]hydroxylamine
C1=CC(=CC=C1C=NO)[N+](=O)[O-]
Melting Point 126°C to 130°C
Quantity 5g
UN Number UN2811
Beilstein 973581
Formula Weight 166.14
Percent Purity 98%
Chemical Name or Material 4-Nitrobenzaldoxime

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H332-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

EINECSNumber : 214-445-5

RTECSNumber : CU7450000

TSCA : No

Recommended Storage : Ambient temperatures