Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzenesulfonyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
98-74-8
C6H4ClNO4S
221.62
MFCD00007445
JXRGUPLJCCDGKG-UHFFFAOYSA-N
4-nitrobenzenesulfonylchloride,4-nitrobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-nitro,4-nitrobenzenesulphonyl chloride,4-nitrophenylsulfonyl chloride,p-nitrophenylsulfonyl chloride,4-nitrobenzenesulfonic acid chloride,p-nitrobenzenesulfonyl chloride,4-nitrobenzenesulphonylchloride,4-nitro-benzenesulfonyl chloride
7404
4-nitrobenzenesulfonyl chloride
C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)Cl
98-74-8
C6H4ClNO4S
221.62
MFCD00007445
JXRGUPLJCCDGKG-UHFFFAOYSA-N
4-nitrobenzenesulfonylchloride,4-nitrobenzene-1-sulfonyl chloride,benzenesulfonyl chloride, 4-nitro,4-nitrobenzenesulphonyl chloride,4-nitrophenylsulfonyl chloride,p-nitrophenylsulfonyl chloride,4-nitrobenzenesulfonic acid chloride,p-nitrobenzenesulfonyl chloride,4-nitrobenzenesulphonylchloride,4-nitro-benzenesulfonyl chloride
7404
4-nitrobenzenesulfonyl chloride
C1=CC(=CC=C1[N+](=O)[O-])S(=O)(=O)Cl
Boiling Point 143°C to 144°C (2.0 hPa)
Packaging Glass bottle
Color Yellow to Brown
Melting Point 75°C to 80°C
Quantity 25g
Infrared Spectrum Authentic
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 94% min. (GC)
Linear Formula SO2ClC6H4NO2
Beilstein 11, 72
Solubility Information Solubility in water: insoluble
Formula Weight 221.62
Physical Form Powder
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzenesulfonyl chloride, 95%
Show More Show Less

GHS H Statement
Harmful to aquatic life with long lasting effects.
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 202-697-9

TSCA : TSCA