Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Nitrobenzhydrazide, 98+%

Click to view available options
Quantity:
10g
50g
250g
636-97-5
C7H7N3O3
181.151
MFCD00007604
FKZXYJYTUSGIQE-UHFFFAOYSA-N
4-nitrobenzhydrazide,p-nitrobenzhydrazide,4-nitrobenzoylhydrazine,p-nitrobenzohydrazide,p-nitrobenzoylhydrazide,p-nitrobenzoic hydrazide,p-nitrobenzoic acid hydrazide,p-nitrobenzoyl hydrazine,4-nitrobenzoic acid hydrazide,4-nitrobenzoyl hydrazide
3693744
CHEBI:66928
4-nitrobenzohydrazide
C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-]
636-97-5
C7H7N3O3
181.151
MFCD00007604
FKZXYJYTUSGIQE-UHFFFAOYSA-N
4-nitrobenzhydrazide,p-nitrobenzhydrazide,4-nitrobenzoylhydrazine,p-nitrobenzohydrazide,p-nitrobenzoylhydrazide,p-nitrobenzoic hydrazide,p-nitrobenzoic acid hydrazide,p-nitrobenzoyl hydrazine,4-nitrobenzoic acid hydrazide,4-nitrobenzoyl hydrazide
3693744
CHEBI:66928
4-nitrobenzohydrazide
C1=CC(=CC=C1C(=O)NN)[N+](=O)[O-]
Melting Point ~216°C (decomposition)
Quantity 250g
Beilstein 519882
Formula Weight 181.15
Percent Purity ≥98%
Chemical Name or Material 4-Nitrobenzhydrazide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 211-271-1

RTECSNumber : DH5670000

TSCA : Yes

Recommended Storage : Ambient temperatures