Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzoic Acid 99+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
1kg
Packaging:
Plastic bottle
62-23-7
C7H5NO4
167.12
MFCD00007352
OTLNPYWUJOZPPA-UHFFFAOYSA-N
4-nitrobenzoic acid,p-nitrobenzoic acid,benzoic acid, 4-nitro,nitrodracylic acid,4-nitrodracylic acid,1-carboxy-4-nitrobenzene,benzoic acid, p-nitro,p-nitrobenzoicacid,p-nitrobenzenecarboxylic acid,kyselina p-nitrobenzoova
6108
CHEBI:262350
C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]
62-23-7
C7H5NO4
167.12
MFCD00007352
OTLNPYWUJOZPPA-UHFFFAOYSA-N
4-nitrobenzoic acid,p-nitrobenzoic acid,benzoic acid, 4-nitro,nitrodracylic acid,4-nitrodracylic acid,1-carboxy-4-nitrobenzene,benzoic acid, p-nitro,p-nitrobenzoicacid,p-nitrobenzenecarboxylic acid,kyselina p-nitrobenzoova
6108
CHEBI:262350
C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]
Infrared Spectrum Authentic
Color Yellow
Melting Point 239.0°C to 242.0°C
Packaging Plastic bottle
Quantity 100g
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula O2NC6H4CO2H
Beilstein 09,389
Merck Index 14,6589
Solubility Information Solubility in water: insoluble. Other solubilities: 1g/110mL alcohol- 1g/45mL ether,1g/12mL methanol- 1g/20mL acetone,1g/150mL chloroform,slightly soluble in benzene and carbon disulfide
Formula Weight 167.12
Physical Form Crystalline Powder
Percent Purity ≥99%
Chemical Name or Material 4-Nitrobenzoic acid, 99+%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 200-526-2

RTECSNumber : DH5075000

TSCA : TSCA