Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzonitrile 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
619-72-7
C7H4N2O2
148.12
MFCD00007279
NKJIFDNZPGLLSH-UHFFFAOYSA-N
benzonitrile, 4-nitro,4-cyanonitrobenzene,p-nitrobenzonitrile,p-cyanonitrobenzene,benzonitrile, p-nitro,4-nitrobenzenecarbonitrile,1-nitro-4-cyanobenzene,4-nitro-benzonitrile,ccris 2328,4-nitrobenzonitril
12090
4-nitrobenzonitrile
C1=CC(=CC=C1C#N)[N+](=O)[O-]
619-72-7
C7H4N2O2
148.12
MFCD00007279
NKJIFDNZPGLLSH-UHFFFAOYSA-N
benzonitrile, 4-nitro,4-cyanonitrobenzene,p-nitrobenzonitrile,p-cyanonitrobenzene,benzonitrile, p-nitro,4-nitrobenzenecarbonitrile,1-nitro-4-cyanobenzene,4-nitro-benzonitrile,ccris 2328,4-nitrobenzonitril
12090
4-nitrobenzonitrile
C1=CC(=CC=C1C#N)[N+](=O)[O-]
Quantity 25g
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
Melting Point 144.0°C to 149.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula O2NC6H4CN
Beilstein 09,397
Solubility Information Solubility in water: sparingly soluble
Formula Weight 148.12
Physical Form Fine Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzonitrile, 97%
Show More Show Less

GHS H Statement:
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-610-0

RTECSNumber : DI4903500

TSCA : TSCA