Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzophenone +99%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1144-74-7
C13H9NO3
227.219
MFCD00007354
ZYMCBJWUWHHVRX-UHFFFAOYSA-N
4-nitrobenzophenone,p-nitrobenzophenone,4-nitrophenyl phenyl methanone,benzophenone, 4-nitro,methanone, 4-nitrophenyl phenyl,4-nitrophenyl phenyl ketone,4-nitrophenyl-phenylmethanone,4-nitrophenyl-phenyl-methanone,4-nitro-benzophenone
70839
(4-nitrophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]
1144-74-7
C13H9NO3
227.219
MFCD00007354
ZYMCBJWUWHHVRX-UHFFFAOYSA-N
4-nitrobenzophenone,p-nitrobenzophenone,4-nitrophenyl phenyl methanone,benzophenone, 4-nitro,methanone, 4-nitrophenyl phenyl,4-nitrophenyl phenyl ketone,4-nitrophenyl-phenylmethanone,4-nitrophenyl-phenyl-methanone,4-nitro-benzophenone
70839
(4-nitrophenyl)-phenylmethanone
C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)[N+](=O)[O-]
Packaging Glass bottle
Melting Point 136°C to 138°C
Color Yellow to Brown
Quantity 5g
Infrared Spectrum Authentic
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula O2NC6H4COC6H5
Beilstein 07, 426
Formula Weight 227.22
Physical Form Fine Crystalline Powder
Percent Purity ≥99.0%
Chemical Name or Material 4-Nitrobenzophenone, 99+%
Show More Show Less

EINECSNumber : 214-542-2

TSCA : TSCA