Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzotrifluoride, 96%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
402-54-0
C7H4F3NO2
191.11
MFCD00007358
XKYLCLMYQDFGKO-UHFFFAOYSA-N
4-nitrobenzotrifluoride,1-nitro-4-trifluoromethyl benzene,p-nitrobenzotrifluoride,benzene, 1-nitro-4-trifluoromethyl,4-nitro-alpha,alpha,alpha-trifluorotoluene,4-trifluoromethyl nitrobenzene,4-nitro-a,a,a-trifluorotoluene,p-nitro-trifluoromethylphenol,p-trifluoromethylnitrobenzene,4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene
9821
1-nitro-4-(trifluoromethyl)benzene
C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-]
402-54-0
C7H4F3NO2
191.11
MFCD00007358
XKYLCLMYQDFGKO-UHFFFAOYSA-N
4-nitrobenzotrifluoride,1-nitro-4-trifluoromethyl benzene,p-nitrobenzotrifluoride,benzene, 1-nitro-4-trifluoromethyl,4-nitro-alpha,alpha,alpha-trifluorotoluene,4-trifluoromethyl nitrobenzene,4-nitro-a,a,a-trifluorotoluene,p-nitro-trifluoromethylphenol,p-trifluoromethylnitrobenzene,4-nitro-.alpha.,.alpha.,.alpha.-trifluorotoluene
9821
1-nitro-4-(trifluoromethyl)benzene
C1=CC(=CC=C1C(F)(F)F)[N+](=O)[O-]
Infrared Spectrum Authentic
Packaging Glass bottle
Flash Point 88°C
Quantity 5g
Color White to Yellow
Melting Point 37.0°C to 42.0°C
Boiling Point 81.0°C to 83.0°C (10.0mmHg)
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 95% min. (GC)
Linear Formula O2NC6H4CF3
Beilstein 05, II, 251
Formula Weight 191.11
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Percent Purity 96%
Chemical Name or Material 4-Nitrobenzotrifluoride, 96%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated cl

GHS Signal Word: Danger

EINECSNumber : 206-948-3