Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl Alcohol 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
619-73-8
C7H7NO3
153.14
MFCD00007376
JKTYGPATCNUWKN-UHFFFAOYSA-N
4-nitrobenzyl alcohol,4-nitrophenyl methanol,p-nitrobenzyl alcohol,benzenemethanol, 4-nitro,4-nitrobenzenemethanol,p-hydroxymethyl nitrobenzene,benzyl alcohol, p-nitro,paranitrobenzyl alcohol,4-nitrobenzylalcohol,unii-86btj68y9m
69275
CHEBI:41214
(4-nitrophenyl)methanol
C1=CC(=CC=C1CO)[N+](=O)[O-]
619-73-8
C7H7NO3
153.14
MFCD00007376
JKTYGPATCNUWKN-UHFFFAOYSA-N
4-nitrobenzyl alcohol,4-nitrophenyl methanol,p-nitrobenzyl alcohol,benzenemethanol, 4-nitro,4-nitrobenzenemethanol,p-hydroxymethyl nitrobenzene,benzyl alcohol, p-nitro,paranitrobenzyl alcohol,4-nitrobenzylalcohol,unii-86btj68y9m
69275
CHEBI:41214
(4-nitrophenyl)methanol
C1=CC(=CC=C1CO)[N+](=O)[O-]
Melting Point 92.0°C to 95.0°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 100g
Boiling Point 185.0°C (12.0 mmHg)
Color Yellow
Flash Point 180°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula O2NC6H4CH2OH
Beilstein 06, 450
Solubility Information Solubility in water: 2g/L (20°C)
Formula Weight 153.14
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzyl alcohol, 99%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 210-611-6

RTECSNumber : DP0657100

TSCA : TSCA