Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl bromide, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
100-11-8
C7H6BrNO2
216.03
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
4-nitrobenzyl bromide,1-bromomethyl-4-nitrobenzene,p-nitrobenzyl bromide,nitrobenzyl bromide,alpha-bromo-4-nitrotoluene,benzene, 1-bromomethyl-4-nitro,p-bromomethyl nitrobenzene,4-bromomethyl nitrobenzene,alpha-bromo-p-nitrotoluene,4-nitrobenzylbromide
66011
CHEBI:62822
1-(bromomethyl)-4-nitrobenzene
C1=CC(=CC=C1CBr)[N+](=O)[O-]
100-11-8
C7H6BrNO2
216.03
VOLRSQPSJGXRNJ-UHFFFAOYSA-N
4-nitrobenzyl bromide,1-bromomethyl-4-nitrobenzene,p-nitrobenzyl bromide,nitrobenzyl bromide,alpha-bromo-4-nitrotoluene,benzene, 1-bromomethyl-4-nitro,p-bromomethyl nitrobenzene,4-bromomethyl nitrobenzene,alpha-bromo-p-nitrotoluene,4-nitrobenzylbromide
66011
CHEBI:62822
1-(bromomethyl)-4-nitrobenzene
C1=CC(=CC=C1CBr)[N+](=O)[O-]
Packaging Glass Bottle
Color Yellow to Beige
Infrared Spectrum Authentic
Flash Point >100°C
Quantity 500g
Melting Point 98.0°C to 100.0°C
Assay Percent Range 99%
Linear Formula O2NC6H4CH2Br
Beilstein 05,334
Fieser 01,736; 08,36
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dmf,isopropyl acetate and acetone
Formula Weight 216.03
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzyl bromide
Show More Show Less