Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl chloroformate, 95%, Acros Organics

Click to view available options
Quantity:
5g
25g
4457-32-3
C8H6ClNO4
215.59
MFCD00007375
MHSGOABISYIYKP-UHFFFAOYSA-N
4-nitrobenzyl chloroformate,4-nitrobenzyl carbonochloridate,carbonochloridic acid, 4-nitrophenyl methyl ester,4-nitrobenzyl chlorocarbonate,p-nitrobenzyl chloroformate,4-nitrocarbobenzoxychloride,4-nitrophenyl methyl chloroformate,p-nitrobenzyloxycarbonyl chloride,4-nitrobenzyloxycarbonyl chloride,chloroformic acid p-nitrobenzyl ester
78205
(4-nitrophenyl)methyl carbonochloridate
C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]
4457-32-3
C8H6ClNO4
215.59
MFCD00007375
MHSGOABISYIYKP-UHFFFAOYSA-N
4-nitrobenzyl chloroformate,4-nitrobenzyl carbonochloridate,carbonochloridic acid, 4-nitrophenyl methyl ester,4-nitrobenzyl chlorocarbonate,p-nitrobenzyl chloroformate,4-nitrocarbobenzoxychloride,4-nitrophenyl methyl chloroformate,p-nitrobenzyloxycarbonyl chloride,4-nitrobenzyloxycarbonyl chloride,chloroformic acid p-nitrobenzyl ester
78205
(4-nitrophenyl)methyl carbonochloridate
C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]
Melting Point 32°C
Refractive Index 1.5520 to 1.556
Flash Point 113°C
Packaging Glass Bottle
Quantity 5g
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Linear Formula ClCO2CH2C6H4NO2
Formula Weight 215.59
Percent Purity 95%
Chemical Name or Material 4-Nitrobenzyl chloroformate

GHS H Statement
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye damage.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rins

GHS Signal Word: Danger

EINECSNumber : 224-708-6