Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl triphenylphosphonium bromide, 97%, Acros Organics

Click to view available options
Quantity:
5g
2767-70-6
C25H21BrNO2P
478.326
MFCD00032108
IPJPTPFIJLFWLP-UHFFFAOYSA-M
4-nitrobenzyl triphenylphosphonium bromide,p-nitrobenzyl triphenylphosphonium bromide,phosphonium, p-nitrobenzyl triphenyl-, bromide,4-nitrobenzyl triphenylphosphoniumbromide,4-nitrobenzyl triphenyl phosphonium bromide,4-nitrophenyl methyl triphenylphosphanium bromide,acmc-209gym,phosphonium, 4-nitrophenyl methyl triphenyl-, bromide,wln: wnr d1pr&r&r &e,4-nitrobenzyltriphenylphosphonium bromide
197008
(4-nitrophenyl)methyl-triphenylphosphanium;bromide
C1=CC=C(C=C1)[P+](CC2=CC=C(C=C2)[N+](=O)[O-])(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
2767-70-6
C25H21BrNO2P
478.326
MFCD00032108
IPJPTPFIJLFWLP-UHFFFAOYSA-M
4-nitrobenzyl triphenylphosphonium bromide,p-nitrobenzyl triphenylphosphonium bromide,phosphonium, p-nitrobenzyl triphenyl-, bromide,4-nitrobenzyl triphenylphosphoniumbromide,4-nitrobenzyl triphenyl phosphonium bromide,4-nitrophenyl methyl triphenylphosphanium bromide,acmc-209gym,phosphonium, 4-nitrophenyl methyl triphenyl-, bromide,wln: wnr d1pr&r&r &e,4-nitrobenzyltriphenylphosphonium bromide
197008
(4-nitrophenyl)methyl-triphenylphosphanium;bromide
C1=CC=C(C=C1)[P+](CC2=CC=C(C=C2)[N+](=O)[O-])(C3=CC=CC=C3)C4=CC=CC=C4.[Br-]
for C-C Bond Formation (Olefination)
Infrared Spectrum Authentic
Melting Point 270°C to 275°C
Color White to Yellow
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 480.34
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material 4-Nitrobenzyl triphenylphosphonium bromide, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 220-444-0

RTECSNumber : TA2390000