Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrocatechol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
10g
50g
Packaging:
Glass bottle
3316-09-4
C6H5NO4
155.11
MFCD00007242
XJNPNXSISMKQEX-UHFFFAOYSA-N
4-nitrocatechol,4-nitropyrocatechol,1,2-dihydroxy-4-nitrobenzene,4-nitro-1,2-benzenediol,3,4-dihydroxy-1-nitrobenzene,unii-sw60ng75en,sw60ng75en,1,2-benzenediol, 4-nitro,nitrocatechol mix of isomers,4nc
3505109
CHEBI:16318
4-nitrobenzene-1,2-diol
C1=CC(=C(C=C1[N+](=O)[O-])O)O
3316-09-4
C6H5NO4
155.11
MFCD00007242
XJNPNXSISMKQEX-UHFFFAOYSA-N
4-nitrocatechol,4-nitropyrocatechol,1,2-dihydroxy-4-nitrobenzene,4-nitro-1,2-benzenediol,3,4-dihydroxy-1-nitrobenzene,unii-sw60ng75en,sw60ng75en,1,2-benzenediol, 4-nitro,nitrocatechol mix of isomers,4nc
3505109
CHEBI:16318
4-nitrobenzene-1,2-diol
C1=CC(=C(C=C1[N+](=O)[O-])O)O
Infrared Spectrum Authentic
Melting Point 173°C to 177°C
Color Yellow
Packaging Glass bottle
Quantity 50g
Sulfated Ash 0.1% max.
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 06, 788
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone
Formula Weight 155.11
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 4-Nitrocatechol
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 222-009-