Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrocinnamyl alcohol, 98%, Thermo Scientific™

Click to view available options
Quantity:
1g
5g
25g
1504-63-8
C9H9NO3
179.175
MFCD00017045
LGXXEDSIJZHDBN-OWOJBTEDSA-N
4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
5462894
(E)-3-(4-nitrophenyl)prop-2-en-1-ol
C1=CC(=CC=C1C=CCO)[N+](=O)[O-]
1504-63-8
C9H9NO3
179.175
MFCD00017045
LGXXEDSIJZHDBN-OWOJBTEDSA-N
4-nitrocinnamyl alcohol, 2e-3-4-nitrophenyl prop-2-en-1-ol, e-3-4-nitrophenyl prop-2-en-1-ol, p-nitrocinnamyl alcohol, 3-4-nitrophenyl prop-2-en-1-ol, 2-propen-1-ol, 3-4-nitrophenyl, 2-propen-1-ol, 3-p-nitrophenyl, 3-p-nitrophenyl allyl alcohol, 3-4-nitrophenyl-2-propenol
5462894
(E)-3-(4-nitrophenyl)prop-2-en-1-ol
C1=CC(=CC=C1C=CCO)[N+](=O)[O-]

4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Nitrocinnamyl alcohol is used in the preparation of 4-nitrocinnamyl chloride by reacting with tosyl chloride (TsCl).

Solubility
Soluble in chloroform, dichloromethane and ethyl Acetate.

Notes
Incompatible with oxidizing agents.
Melting Point 126°C to 130°C
Beilstein 1948238
Quantity 25g
Solubility Information Soluble in chloroform,dichloromethane and ethyl Acetate.
Formula Weight 179.18
Percent Purity 98%
Chemical Name or Material 4-Nitrocinnamyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : Yes

Recommended Storage : Ambient temperatures

RUO – Research Use Only