Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrohippuric Acid 98%, Acros Organics

Click to view available options
Quantity:
50g
2645-07-0
C9H8N2O5
224.172
MFCD00007349
XCMUCRMMVDSVEL-UHFFFAOYSA-N
4-nitrohippuric acid,p-nitrohippuric acid,glycine, n-4-nitrobenzoyl,n-4-nitrobenzoyl glycine,n-4-nitrobenzoylglycine,p-nitrobenzoylglycine,2-4-nitrobenzamido acetic acid,hippuric acid, p-nitro,n-p-nitrobenzoyl glycine,unii-9hiu46t98m
17549
2-[(4-nitrobenzoyl)amino]acetic acid
C1=CC(=CC=C1C(=O)NCC(=O)O)[N+](=O)[O-]
2645-07-0
C9H8N2O5
224.172
MFCD00007349
XCMUCRMMVDSVEL-UHFFFAOYSA-N
4-nitrohippuric acid,p-nitrohippuric acid,glycine, n-4-nitrobenzoyl,n-4-nitrobenzoyl glycine,n-4-nitrobenzoylglycine,p-nitrobenzoylglycine,2-4-nitrobenzamido acetic acid,hippuric acid, p-nitro,n-p-nitrobenzoyl glycine,unii-9hiu46t98m
17549
2-[(4-nitrobenzoyl)amino]acetic acid
C1=CC(=CC=C1C(=O)NCC(=O)O)[N+](=O)[O-]
Infrared Spectrum Authentic
Melting Point 126°C to 130°C
Quantity 50g
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Linear Formula O2NC6H4CONHCH2CO2H
Beilstein 09, 395
Formula Weight 224.17
Physical Form Granules
Percent Purity ≥97.5%
Chemical Name or Material 4-Nitrohippuric acid, 98%
Show More Show Less