Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenethyl Bromide 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5339-26-4
C8H8BrNO2
230.06
MFCD00007386
NTURQZFFJDCTMZ-UHFFFAOYSA-N
4-nitrophenethyl bromide,1-2-bromoethyl-4-nitrobenzene,benzene, 1-2-bromoethyl-4-nitro,4-nitrophenethylbromide,p-nitrophenethyl bromide,2-4-nitrophenyl ethyl bromide,4-2-bromoethyl nitrobenzene,4-nitrophenylethyl bromide,.beta.-p-nitrophenyl ethyl bromide,phenethylbromide4nitro
79266
1-(2-bromoethyl)-4-nitrobenzene
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
5339-26-4
C8H8BrNO2
230.06
MFCD00007386
NTURQZFFJDCTMZ-UHFFFAOYSA-N
4-nitrophenethyl bromide,1-2-bromoethyl-4-nitrobenzene,benzene, 1-2-bromoethyl-4-nitro,4-nitrophenethylbromide,p-nitrophenethyl bromide,2-4-nitrophenyl ethyl bromide,4-2-bromoethyl nitrobenzene,4-nitrophenylethyl bromide,.beta.-p-nitrophenyl ethyl bromide,phenethylbromide4nitro
79266
1-(2-bromoethyl)-4-nitrobenzene
C1=CC(=CC=C1CCBr)[N+](=O)[O-]
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
Color Yellow
Melting Point 67°C to 70°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula O2NC6H4CH2CH2Br
Formula Weight 230.06
Physical Form Adhering Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Nitrophenethyl bromide, 98%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-271-7