Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrophenyl 2-(trimethylsilyl)ethyl carbonate, 95%, Acros Organics

Click to view available options
Quantity:
25g
Packaging:
Glass Bottle
80149-80-0
C12H17NO5Si
283.355
ZAQWGGKIMQIVGM-UHFFFAOYSA-N
4-nitrophenyl 2-trimethylsilyl ethyl carbonate,teoc-onp,2-trimethylsilyl ethyl p-nitrophenyl carbonate,teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate,4-nitrophenyl 2-trimethylsilylethyl carbonate,2-trimethylsilyl ethyl 4-nitrophenyl carbonate,4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene,carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester,ambotzrl-8204
3086116
(4-nitrophenyl) 2-trimethylsilylethyl carbonate
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
80149-80-0
C12H17NO5Si
283.355
ZAQWGGKIMQIVGM-UHFFFAOYSA-N
4-nitrophenyl 2-trimethylsilyl ethyl carbonate,teoc-onp,2-trimethylsilyl ethyl p-nitrophenyl carbonate,teoc-onp;4-nitrophenyl-2-trimethylsilyl ethyl carbonate,4-nitrophenyl 2-trimethylsilylethyl carbonate,2-trimethylsilyl ethyl 4-nitrophenyl carbonate,4-2-trimethylsilyl ethoxycarbonyloxy nitrobenzene,carbonic acid, 4-nitrophenyl 2-trimethylsilyl ethyl ester,ambotzrl-8204
3086116
(4-nitrophenyl) 2-trimethylsilylethyl carbonate
C[Si](C)(C)CCOC(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
Packaging Glass Bottle
Infrared Spectrum Authentic
Melting Point 35°C to 40°C
Quantity 25g
Color Yellow
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 283.36
Physical Form Crystalline Powder
Percent Purity ≥94.0%
Chemical Name or Material 4-Nitrophenyl 2-(trimethylsilyl)ethylecarbonate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.